missing translation for 'onlineSavingsMsg'
Learn More

(1-Methyl-2-piperidnyl)methanamine, 97%, Thermo Scientific™

Product Code. 10450584
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 5298-72-6
Molecular Formula C7H16N2
Molecular Weight (g/mol) 128.22
MDL Number MFCD05022432
InChI Key PPUMJZMVFCLQBI-UHFFFAOYNA-N
PubChem CID 2794677
SMILES CN1CCCCC1CN

Specifications

Density 0.9102g/mL
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Synonym 1-methylpiperidin-2-yl methanamine, 1-methyl-2-piperidinyl methanamine, 2-piperidinemethanamine, 1-methyl, 1-1-methylpiperidin-2-yl methanamine, c-1-methyl-piperidin-2-yl-methylamine, 1-methyl-2-piperidnyl methanamine, 1-methyl-2-piperidyl methylamine, pubchem8033, 1-methyl-2-aminomethylpiperidine, 2-piperidinemethanamine,1-methyl
IUPAC Name (1-methylpiperidin-2-yl)methanamine
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material (1-Methyl-2-piperidinyl)methanamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.