missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Methyl-2-piperidnyl)methanamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Density | 0.9102g/mL |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 1-methylpiperidin-2-yl methanamine, 1-methyl-2-piperidinyl methanamine, 2-piperidinemethanamine, 1-methyl, 1-1-methylpiperidin-2-yl methanamine, c-1-methyl-piperidin-2-yl-methylamine, 1-methyl-2-piperidnyl methanamine, 1-methyl-2-piperidyl methylamine, pubchem8033, 1-methyl-2-aminomethylpiperidine, 2-piperidinemethanamine,1-methyl |
IUPAC Name | (1-methylpiperidin-2-yl)methanamine |
Formula Weight | 128.22 |
Percent Purity | 97% |
Chemical Name or Material | (1-Methyl-2-piperidinyl)methanamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction