missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-2-pyrrolidinone, ACS Reagent, Honeywell
Click to view available options
Quantity:
500 mL
1 L
2.5 L
Specifications
Specifications
Density | 1.028g/mL at 25°C |
Quantity | 2.5 L |
Explosion Limit | 0.095 |
Flash Point | 91°C (195.8°F) |
Ignition Point | 518°F |
Vapor Pressure | 0.29 mmHg (20°C), 0.99 mmHg (40°C) |
Beilstein | 106420 |
Formula Weight | 99.13g/mol |
Percent Purity | ≥99% |
Grade | ACS Reagent |
Show More |
Safety and Handling
Product Identifier
- 1-Methyl-2-pyrrolidinone
- Danger
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H360D-May damage the unborn child.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : N-Methyl-2-pyrrolidone
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'