missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-2-pyrrolidinone, ACS Reagent, Honeywell

Product Code. 15695540
Click to view available options
Quantity:
500 mL
1 L
2.5 L
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 872-50-4
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.133
MDL Number MFCD00003193
InChI Key SECXISVLQFMRJM-UHFFFAOYSA-N
Synonym 1-methyl-2-pyrrolidinone, n-methyl-2-pyrrolidone, n-methylpyrrolidone, 1-methyl-2-pyrrolidone, methylpyrrolidone, m-pyrol, n-methyl-2-pyrrolidinone, n-methylpyrrolidinone, 1-methylpyrrolidinone, 2-pyrrolidinone, 1-methyl
PubChem CID 13387
ChEBI CHEBI:7307
IUPAC Name 1-methylpyrrolidin-2-one
SMILES CN1CCCC1=O

Specifications

Density 1.028g/mL at 25°C
Quantity 500 mL
Explosion Limit 0.095
Flash Point 91°C (195.8°F)
Ignition Point 518°F
Vapor Pressure 0.29 mmHg (20°C), 0.99 mmHg (40°C)
Beilstein 106420
Formula Weight 99.13g/mol
Percent Purity ≥99%
Grade ACS Reagent
Chemical Name or Material 1-Methyl-2-pyrrolidinone
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'