missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-2-(tri-n-butylstannyl)pyrrole, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
It is employed as intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is employed as intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is employed as intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 1.122 |
Flash Point | 98°C (208°F) |
Refractive Index | 1.511 |
Quantity | 1 g |
UN Number | UN2788 |
Solubility Information | Not miscible or difficult to mix. |
Formula Weight | 370.15 |
Percent Purity | 98% |
Chemical Name or Material | 1-Methyl-2-(tri-n-butylstannyl)pyrrole |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'