missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-indoleacetic acid, 98%

Product Code. p-4228803 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11996681

Brand: Thermo Scientific Acros 439240010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1912-48-7 | C11H11NO2 | 189.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1912-48-7
Molecular Formula C11H11NO2
Molecular Weight (g/mol) 189.21
InChI Key NAIPEFIYIQFVFC-UHFFFAOYSA-N
Synonym 1-methyl-3-indoleacetic acid, 2-1-methyl-1h-indol-3-yl acetic acid, 1h-indole-3-acetic acid, 1-methyl, 2-1-methylindol-3-yl acetic acid, 1-methylindole-3-acetic acid, 1-methyl-3-indolyl acetic acid, 1-methylindol-3-yl acetic acid, 1-methyl-1h-indole-3-acetic acid, 1-methyl-3-indoleaceticacid, 1-methyl-1 h-indol-3-yl acetic acid
PubChem CID 254166
IUPAC Name 2-(1-methylindol-3-yl)acetic acid
SMILES CN1C=C(C2=CC=CC=C21)CC(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 125.0°C to 126.5°C
Color Tan to White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 189.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 1-Methyl-3-indoleacetic acid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Methyl-3-indoleacetic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.