missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonyl chloride, 97+%, Thermo Scientific™

Product Code. 10709615
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 126674-98-4
Molecular Formula C6H4ClF3N2O
Molecular Weight (g/mol) 212.556
InChI Key KFKVECZQALNWSR-UHFFFAOYSA-N
Synonym 1-methyl-3-trifluoromethyl-1h-pyrazole-4-carbonyl chloride, 1-methyl-3-trifluoromethyl pyrazole-4-carbonyl chloride, 1h-pyrazole-4-carbonylchloride, 1-methyl-3-trifluoromethyl, acmc-20ms45, 3-trifluoromethyl-1-methyl-1h-pyrazole-4-carbonyl chloride, 1-methyl-3-trifluoromethylpyrazole-4-carboxylic acid chloride, 1-methyl-4-trifluoromethyl-1h-pyrazole-3-carbonyl chloride, 1-methyl-3-trifluoromethyl-1h-pyrazole-4-carboxylic acid chloride, 1h-pyrazole-4-carbonyl chloride, 1-methyl-3-trifluoromethyl
PubChem CID 2794581
IUPAC Name 1-methyl-3-(trifluoromethyl)pyrazole-4-carbonyl chloride
SMILES CN1C=C(C(=N1)C(F)(F)F)C(=O)Cl

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 212.56
Percent Purity 95%
Chemical Name or Material 1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-carbonyl chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'