missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-(3-nitrophenyl)piperazine, 97%, Thermo Scientific™

Product Code. 10183714
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 148546-97-8
Molecular Formula C11H15N3O2
Molecular Weight (g/mol) 221.26
MDL Number MFCD08435858
InChI Key IIRKKCDXJIXWHI-UHFFFAOYSA-N
Synonym 1-methyl-4-3-nitrophenyl piperazine, 4-methyl-1-3-nitrophenyl piperazine, piperazine,1-methyl-4-3-nitrophenyl, zlchem 196, pubchem18685, acmc-209d0u, 1-3-nitrophenyl 4-methylpiperazine, 1-3-nitrophenyl-4-methylpiperazine, 1-methyl-4-3-nitrophenyl-piperazine, 1-methyl-4-3-nitro-phenyl-piperazine
PubChem CID 12152161
IUPAC Name 1-methyl-4-(3-nitrophenyl)piperazine
SMILES CN1CCN(CC1)C1=CC(=CC=C1)[N+]([O-])=O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 221.26
Percent Purity 97%
Chemical Name or Material 1-Methyl-4-(3-nitrophenyl)piperazine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'