missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-Methyl-4-piperidnyl)methanamine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 128.22 |
Percent Purity | 97% |
Chemical Name or Material | (1-Methyl-4-piperidinyl)methanamine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction