missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(1-methyl-5-phenyl-1H-pyrazol-4-yl)methanol, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
Specifications
Quantity | 250 mg |
IUPAC Name | (1-methyl-5-phenyl-1H-pyrazol-4-yl)methanol |
Formula Weight | 188.23 |
Percent Purity | 97% |
Chemical Name or Material | (1-methyl-5-phenyl-1H-pyrazol-4-yl)methanol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction