missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-5-phenyl-1H-pyrazole-3-carboxylic acid, 97%, Thermo Scientific™

Catalog Number 11320637
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
10199-53-8
C11H10N2O2
202.21
MFCD08271933
ZDUDGEACIDDEPL-UHFFFAOYSA-N
1-methyl-5-phenyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 1-methyl-5-phenyl, 1h-pyrazole-3-carboxylic acid, 1-methyl-5-phenyl, 3-carboxy-1-methyl-5-phenyl-1h-pyrazole, 1-methyl-5-phenyl pyrazole-3-carboxylic acid, 3-carboxy-1-methyl-1h-pyrazol-5-yl benzene, 1-methyl-5-phenyl-1h-3-pyrazolecarboxylic acid
7537619
1-methyl-5-phenylpyrazole-3-carboxylic acid
CN1N=C(C=C1C1=CC=CC=C1)C(O)=O
This item is not returnable. View return policy
10199-53-8
C11H10N2O2
202.21
MFCD08271933
ZDUDGEACIDDEPL-UHFFFAOYSA-N
1-methyl-5-phenyl-1h-pyrazole-3-carboxylic acid, 1h-pyrazole-3-carboxylicacid, 1-methyl-5-phenyl, 1h-pyrazole-3-carboxylic acid, 1-methyl-5-phenyl, 3-carboxy-1-methyl-5-phenyl-1h-pyrazole, 1-methyl-5-phenyl pyrazole-3-carboxylic acid, 3-carboxy-1-methyl-1h-pyrazol-5-yl benzene, 1-methyl-5-phenyl-1h-3-pyrazolecarboxylic acid
7537619
1-methyl-5-phenylpyrazole-3-carboxylic acid
CN1N=C(C=C1C1=CC=CC=C1)C(O)=O
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 202.21
Percent Purity 97%
Chemical Name or Material 1-Methyl-5-phenyl-1H-pyrazole-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'