missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methyl-5-phenyl-1H-pyrazole-4-carbaldehyde, Thermo Scientific™
£70.20 - £615.00
Chemical Identifiers
CAS | 154927-01-2 |
---|---|
Molecular Formula | C11H10N2O |
Molecular Weight (g/mol) | 186.214 |
InChI Key | DLMVCAXLQMJYHP-UHFFFAOYSA-N |
Synonym | 1-methyl-5-phenyl-1h-pyrazole-4-carbaldehyde, 1-methyl-5-phenyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde,1-methyl-5-phenyl |
PubChem CID | 2795475 |
IUPAC Name | 1-methyl-5-phenylpyrazole-4-carbaldehyde |
SMILES | CN1C(=C(C=N1)C=O)C2=CC=CC=C2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10758643
|
Thermo Scientific Maybridge
CC28104CB |
250 mg | Amber Glass Bottle |
£70.20
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10137672
|
Thermo Scientific Maybridge
CC28104DA |
1 g | Amber Glass Bottle |
£242.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10396234
|
Thermo Scientific Maybridge
CC28104DE |
5 g | Amber Glass Bottle |
£615.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
154927-01-2 | |
186.214 | |
1-methyl-5-phenyl-1h-pyrazole-4-carbaldehyde, 1-methyl-5-phenyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde,1-methyl-5-phenyl | |
1-methyl-5-phenylpyrazole-4-carbaldehyde |
C11H10N2O | |
DLMVCAXLQMJYHP-UHFFFAOYSA-N | |
2795475 | |
CN1C(=C(C=N1)C=O)C2=CC=CC=C2 |
Specifications
154927-01-2 | |
C11H10N2O | |
DLMVCAXLQMJYHP-UHFFFAOYSA-N | |
1-methyl-5-phenylpyrazole-4-carbaldehyde | |
2795475 | |
97% |
97% | |
1-methyl-5-phenyl-1h-pyrazole-4-carbaldehyde, 1-methyl-5-phenyl-1h-pyrazole-4-carboxaldehyde, 1h-pyrazole-4-carboxaldehyde,1-methyl-5-phenyl | |
CN1C(=C(C=N1)C=O)C2=CC=CC=C2 | |
186.214 | |
186.21 | |
1-Methyl-5-phenyl-1H-pyrazole-4-carbaldehyde |