missing translation for 'onlineSavingsMsg'
Learn More

1-Methylcyclopentanol, 98%, Thermo Scientific Chemicals

£50.60 - £531.00

Chemical Identifiers

CAS 1462-03-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.161
MDL Number MFCD00001364
InChI Key CAKWRXVKWGUISE-UHFFFAOYSA-N
PubChem CID 73830
IUPAC Name 1-methylcyclopentan-1-ol
SMILES CC1(CCCC1)O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11456446
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B24744.06
5 g
£50.60
5g
Please to purchase this item. Need a web account? Register with us today!
11466446
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B24744.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11476446
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B24744.22
100 g
£531.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1462-03-9
100.161
CAKWRXVKWGUISE-UHFFFAOYSA-N
1-methylcyclopentan-1-ol
C6H12O
MFCD00001364
73830
CC1(CCCC1)O

Specifications

1462-03-9
0.904
40°C (105°F)
MFCD00001364
1-methylcyclopentanol, 1-methyl cyclopentanol, cyclopentanol, 1-methyl, 1-methyl-cyclopentanol, 1-hydroxy-1-methylcyclopentane, cyclopentanol, methyl, 1-methyl-1-cyclopentanol, 1-methylcyclopentanolc, cyclopentanol,1-methyl
CC1(CCCC1)O
100.161
100.16
1-Methylcyclopentanol
36°C to 37°C
135°C to 136°C
C6H12O
UN1325
CAKWRXVKWGUISE-UHFFFAOYSA-N
1-methylcyclopentan-1-ol
73830
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P240-P241-P280-P370+P378q

H228

missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III

missing translation for 'einecsNumber' : 215-963-4

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only