missing translation for 'onlineSavingsMsg'
Learn More
1-Methylpiperazine, 99%, Thermo Scientific Chemicals Product Code.: 10417940

1-Methylpiperazine, 99%, Thermo Scientific Chemicals

Product Code. 10417940
5 g, Glass bottle
Click to view available options
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Metal Drum
This item is not returnable. View return policy

Product Code. 10417940

Brand: Thermo Scientific Acros 156910050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-01-3
Molecular Formula C5H12N2
Molecular Weight (g/mol) 100.16
InChI Key PVOAHINGSUIXLS-UHFFFAOYSA-N
Synonym n-methylpiperazine, piperazine, 1-methyl, 4-methylpiperazine, 1-methyl-piperazine, n-methyl piperazine, 1-methyl piperazine, methylpiperazine, n-methyl-piperazine, n-methylpiperazin, 1-methylpyperazine
PubChem CID 53167
IUPAC Name 1-methylpiperazine
SMILES CN1CCNCC1

Specifications

Melting Point -6.0°C
Density 0.9030g/mL
Boiling Point 138.0°C
Flash Point 42°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4645 to 1.4665
Quantity 5 g
Specific Gravity 0.903
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol and aceton
Viscosity 4 mPa.s (20°C)
Formula Weight 100.16
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Methylpiperazine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0443711 Certificate TypeCertificate of Analysis Date25/01/2025 Catalog Number156910050,156915000,156910025,156911000
Lot NumberA0384095 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number
Lot NumberA0404170 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number
Lot NumberA0443711 Certificate TypeCertificate of Analysis Date30/06/2022 Catalog Number
Lot NumberA0364473 Certificate TypeCertificate of Analysis Date05/10/2020 Catalog Number
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Methylpiperazine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Methylpiperazine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.