missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpiperidine, 99%, Thermo Scientific Chemicals

£23.10 - £146.00

Chemical Identifiers

CAS 626-67-5
Molecular Formula C6H14ClN
Molecular Weight (g/mol) 135.64
MDL Number MFCD00006491
InChI Key QDUXDCXILAPLAG-UHFFFAOYSA-N
Synonym n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
PubChem CID 12291
IUPAC Name 1-methylpiperidine
SMILES [H+].[Cl-].CN1CCCCC1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11332136
View Documents
Thermo Scientific Alfa Aesar
L03398.09
10 g
£23.10
10g
Please to purchase this item. Need a web account? Register with us today!
11342136
View Documents
Thermo Scientific Alfa Aesar
L03398.22
100 g
£39.10
100g
Please to purchase this item. Need a web account? Register with us today!
11352136
View Documents
Thermo Scientific Alfa Aesar
L03398.36
500 g
£146.00
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-Methylpiperidine is a reactant for sp3 C-H Bond activation with ruthenium(II) catalysts and C(3)-alkylation of cyclic amines, One-pot synthesis of Z-cinnamic acids. It is used in the synthesis of Unsymmetrical ureas, Antibacterial imidazolium, pyrrolidinium, and piperidinium salts, C1-C16 segment of goniodomin A via palladium-catalyzed organostannane thioester coupling.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Methylpiperidine is a reactant for sp3 C-H Bond activation with ruthenium(II) catalysts and C(3)-alkylation of cyclic amines, One-pot synthesis of Z-cinnamic acids. It is used in the synthesis of Unsymmetrical ureas, Antibacterial imidazolium, pyrrolidinium, and piperidinium salts, C1-C16 segment of goniodomin A via palladium-catalyzed organostannane thioester coupling.

Solubility
Fully miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with.acids, Acid chlorides, Acid anhydrides, Strong oxidizing agents, Carbon dioxide.
Specifications

Chemical Identifiers

626-67-5
135.64
QDUXDCXILAPLAG-UHFFFAOYSA-N
12291
[H+].[Cl-].CN1CCCCC1
C6H14ClN
MFCD00006491
n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
1-methylpiperidine

Specifications

626-67-5
0.816
105°C to 107°C
Amine-like
1.4378
UN2399
n-methylpiperidine, piperidine, 1-methyl, methylpiperidine, n-methyl piperidine, 1-methyl-piperidine, n-methylpiperidin, methylpiperdine, 1-methylpiperadine, n-methyl-piperidine, 1-methyl piperidine
QDUXDCXILAPLAG-UHFFFAOYSA-N
1-methylpiperidine
12291
99%
-50°C
12
3°C (37°F)
C6H14ClN
MFCD00006491
1073
Fully miscible in water.
[H+].[Cl-].CN1CCCCC1
135.64
99.18
1-Methylpiperidine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H331-H314-H318-H302
Highly flammable liquid and vapor.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H312+H332-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: 1-METHYLPIPERIDINE

missing translation for 'einecsNumber' : 210-959-9

missing translation for 'rtecsNumber' : TN1225000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only