missing translation for 'onlineSavingsMsg'
Learn More

1-Methylpsilocin, Tocris Bioscience™

Product Code. 18015882
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective 5-HT2C agonist

Potent and selective 5-HT2C agonist (IC50 values are 12 and 633 nM for 5-HT2C and 5-HT2A receptors respectively). Displays high affinity for the 5-HT2B receptor (Ki = 38 nM) but acts as an inverse agonist. Active in vivo; inhibits scratching in a mouse model of OCD.

Chemical Identifiers

CAS 1465-16-3
Molecular Formula C13H18N2O
Molecular Weight (g/mol) 218.3
InChI Key MZZRFEIDRWKTKJ-UHFFFAOYSA-N
Synonym 1-methylpsilocin, 3-2-dimethylamino ethyl-1-methylindol-4-ol, 3-2-dimethylamino-ethyl-1-methyl-1h-indol-4-ol, 3-2-dimethylamino ethyl-1-methyl-1h-indol-4-ol, d0r8vt
PubChem CID 44404883
IUPAC Name 3-[2-(dimethylamino)ethyl]-1-methylindol-4-ol
SMILES CN1C=C(C2=C1C=CC=C2O)CCN(C)C

Specifications

Quantity 10 mg
Formula Weight 218.29
Percent Purity >99%
Chemical Name or Material 1-Methylpsilocin
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'