missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Methylpsilocin, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
Potent and selective 5-HT2C agonist (IC50 values are 12 and 633 nM for 5-HT2C and 5-HT2A receptors respectively). Displays high affinity for the 5-HT2B receptor (Ki = 38 nM) but acts as an inverse agonist. Active in vivo; inhibits scratching in a mouse model of OCD.
Specifications
Specifications
Quantity | 50 mg |
Formula Weight | 218.29 |
Percent Purity | >99% |
Chemical Name or Material | 1-Methylpsilocin |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'