missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium bromide, 99%

Product Code. 11352179
Click to view available options
Quantity:
5 g
50 g
Unit Size:
50g
5g
This item is not returnable. View return policy

Product Code. 11352179

Brand: Thermo Scientific Alfa Aesar H27201.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 85100-77-2 | C8H15BrN2 | 219.126 g/mol

It is an ionic liquid used in a Heck reaction. It is also used in assessing the factors responsible for ionic liquid toxicity to aquatic organisms via quantitative structure-property relationship modeling

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an ionic liquid used in a Heck reaction. It is also used in assessing the factors responsible for ionic liquid toxicity to aquatic organisms via quantitative structure-property relationship modeling

Solubility
Miscible in water, methanol and dichloromethane. Immiscible in acetone, toluene, ethyl acetate and diethyl ether.

Notes
Hygroscopic. Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85100-77-2
Molecular Formula C8H15BrN2
Molecular Weight (g/mol) 219.126
MDL Number MFCD03427611
InChI Key KYCQOKLOSUBEJK-UHFFFAOYSA-M
Synonym 1-butyl-3-methylimidazolium bromide, 3-butyl-1-methyl-1h-imidazol-3-ium bromide, bmimbr, 1-n-butyl-3-methylimidazolium bromide, 1-butyl-3-methyl-1h-imidazol-3-ium bromide, bmim br, c4mim bromide, ksc491c7d, 1-butyl-3-methylimidazoliumbromide
PubChem CID 2734236
IUPAC Name 1-butyl-3-methylimidazol-3-ium;bromide
SMILES CCCCN1C=C[N+](=C1)C.[Br-]

Specifications

Melting Point 65°C to 75°C
Quantity 50 g
Sensitivity Hygroscopic
Solubility Information Miscible in water,methanol and dichloromethane. Immiscible in acetone,toluene,ethyl acetate and diethyl ether.
Formula Weight 219.12
Percent Purity 99%
Chemical Name or Material 1-n-Butyl-3-methylimidazolium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.