missing translation for 'onlineSavingsMsg'
Learn More

1-n-Butyl-3-methylimidazolium dicyanamide, 97%, Thermo Scientific Chemicals

Product Code. 15485408
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 448245-52-1
Molecular Formula C10H15N5
Molecular Weight (g/mol) 205.265
MDL Number MFCD07784457
InChI Key ICIVTHOGIQHZRY-UHFFFAOYSA-N
Synonym 1-butyl-3-methylimidazolium dicyanamide, 1-butyl-3-methylimidazolium dicyamide, 1-butyl-3-methylimidazolium; dicyanoazanide, bmim dca, dsstox_cid_27886, dsstox_rid_82639, dsstox_gsid_47910, c4mim; dicyanoazanide, 1-butyl-3-methyl-1h-imidazol-3-ium cyanoimino methylidene azanide, 1-butyl-3-methylimidazolium; cyanoimino methylidene azanide
PubChem CID 11171745
IUPAC Name 1-butyl-3-methylimidazol-3-ium;cyanoiminomethylideneazanide
SMILES CCCCN1C=C[N+](=C1)C.C(=[N-])=NC#N

Specifications

Color Yellow
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 205.26
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-n-Butyl-3-methylimidazolium dicyanamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'