Learn More
1-n-Butyl-3-methylimidazolium methyl sulfate, 99%, Thermo Scientific Chemicals
£30.35 - £170.85
Chemical Identifiers
CAS | 401788-98-5 |
---|---|
Molecular Formula | C9H18N2O4S |
Molecular Weight (g/mol) | 250.31 |
MDL Number | MFCD03095437 |
InChI Key | MEMNKNZDROKJHP-UHFFFAOYSA-M |
Synonym | 1-butyl-3-methylimidazolium methyl sulfate, 1-butyl-3-methylimidazolium methylsulfate, bmimmeso4, 1-n-butyl-3-methylimidazolium methylsulfate, 1-butyl-3-methylimidazolium methyl sulfate 1-, acmc-20aowh, basionics™ lq 02, 1-butyl-3-methylimidazolium methylsul-fate, 1-butyl-3-methylimidazol-3-ium,methyl sulfate, 1-butyl-3-methyl-imidazol-3-ium; methyl sulfate |
PubChem CID | 12095228 |
SMILES | COS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11382909
|
Thermo Scientific Alfa Aesar
H27754.06 |
5 g |
£30.35
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11392909
|
Thermo Scientific Alfa Aesar
H27754.18 |
50 g |
£170.85
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
401788-98-5 | |
250.31 | |
MEMNKNZDROKJHP-UHFFFAOYSA-M | |
12095228 |
C9H18N2O4S | |
MFCD03095437 | |
1-butyl-3-methylimidazolium methyl sulfate, 1-butyl-3-methylimidazolium methylsulfate, bmimmeso4, 1-n-butyl-3-methylimidazolium methylsulfate, 1-butyl-3-methylimidazolium methyl sulfate 1-, acmc-20aowh, basionics™ lq 02, 1-butyl-3-methylimidazolium methylsul-fate, 1-butyl-3-methylimidazol-3-ium,methyl sulfate, 1-butyl-3-methyl-imidazol-3-ium; methyl sulfate | |
COS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1 |
Specifications
401788-98-5 | |
C9H18N2O4S | |
MFCD03095437 | |
1-butyl-3-methylimidazolium methyl sulfate, 1-butyl-3-methylimidazolium methylsulfate, bmimmeso4, 1-n-butyl-3-methylimidazolium methylsulfate, 1-butyl-3-methylimidazolium methyl sulfate 1-, acmc-20aowh, basionics™ lq 02, 1-butyl-3-methylimidazolium methylsul-fate, 1-butyl-3-methylimidazol-3-ium,methyl sulfate, 1-butyl-3-methyl-imidazol-3-ium; methyl sulfate | |
COS([O-])(=O)=O.CCCCN1C=C[N+](C)=C1 | |
250.31 | |
250.32 | |
1-n-Butyl-3-methylimidazolium methyl sulfate |
1.21 | |
1.478 | |
UN3265 | |
MEMNKNZDROKJHP-UHFFFAOYSA-M | |
1-butyl-3-methyl-1H-imidazol-3-ium methyl sulfate | |
12095228 | |
99% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.