missing translation for 'onlineSavingsMsg'
Learn More

1-n-Decyl-3-methylimidazolium tetrafluoroborate, 97%, Thermo Scientific Chemicals

Product Code. 15494846
Click to view available options
Quantity:
5 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is employed as a intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a intermediate for pharmaceutical.

Notes
Moisture Sensitive, store away from moisture. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 244193-56-4
Molecular Formula C14H27BF4N2
Molecular Weight (g/mol) 310.19
MDL Number MFCD09038853
InChI Key QGUMDWFYJYXDTH-UHFFFAOYSA-N
Synonym 1-decyl-3-methylimidazolium tetrafluoroborate, dsstox_cid_27891, dsstox_rid_82644, dsstox_gsid_47915, ksc200g6n, c10mim bf4, 1-decyl-3-me-thylimidazolium tetrafluoroborate
PubChem CID 11461044
IUPAC Name 1-decyl-3-methylimidazol-3-ium;tetrafluoroborate
SMILES F[B-](F)(F)F.CCCCCCCCCCN1C=C[N+](C)=C1

Specifications

Color Yellow
Refractive Index 1.444
Quantity 5 g
Sensitivity Moisture sensitive
Formula Weight 310.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-n-Decyl-3-methylimidazolium tetrafluoroborate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'