missing translation for 'onlineSavingsMsg'
Learn More

1-n-Hexyl-3-methylimidazolium chloride, 98%, Thermo Scientific Chemicals

£23.20 - £102.51

Chemical Identifiers

CAS 171058-17-6
Molecular Formula C10H19N2
Molecular Weight (g/mol) 167.28
MDL Number MFCD03093289
InChI Key RVEJOWGVUQQIIZ-UHFFFAOYSA-N
Synonym 1-hexyl-3-methylimidazolium chloride, hmimcl, 1-hexyl-3-methyl-1h-imidazol-3-ium chloride, 1-hexyl-3-methylimidazol-3-ium chloride, hmim cl, acmc-209e2c, dsstox_cid_27924, dsstox_rid_82677, dsstox_gsid_47948, ksc491c7f
PubChem CID 2734163
ChEBI CHEBI:61340
IUPAC Name 1-hexyl-3-methylimidazol-3-ium;chloride
SMILES CCCCCC[N+]1=CN(C)C=C1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11446330
View Documents
Thermo Scientific Alfa Aesar
H27178.06
5 g
£23.20
5g
Please to purchase this item. Need a web account? Register with us today!
11456330
View Documents
Thermo Scientific Alfa Aesar
H27178.18
50 g
£102.51
50g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

1-n-Hexyl-3-methylimidazolium chloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-n-Hexyl-3-methylimidazolium chloride, is used as an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents, strong bases.
Specifications

Chemical Identifiers

171058-17-6
167.28
RVEJOWGVUQQIIZ-UHFFFAOYSA-N
2734163
1-hexyl-3-methylimidazol-3-ium;chloride
C10H19N2
MFCD03093289
1-hexyl-3-methylimidazolium chloride, hmimcl, 1-hexyl-3-methyl-1h-imidazol-3-ium chloride, 1-hexyl-3-methylimidazol-3-ium chloride, hmim cl, acmc-209e2c, dsstox_cid_27924, dsstox_rid_82677, dsstox_gsid_47948, ksc491c7f
CHEBI:61340
CCCCCC[N+]1=CN(C)C=C1

Specifications

171058-17-6
1.5165
1-hexyl-3-methylimidazolium chloride, hmimcl, 1-hexyl-3-methyl-1h-imidazol-3-ium chloride, 1-hexyl-3-methylimidazol-3-ium chloride, hmim cl, acmc-209e2c, dsstox_cid_27924, dsstox_rid_82677, dsstox_gsid_47948, ksc491c7f
RVEJOWGVUQQIIZ-UHFFFAOYSA-N
1-hexyl-3-methylimidazol-3-ium;chloride
2734163
202.72
1-n-Hexyl-3-methylimidazolium chloride
C10H19N2
MFCD03093289
Insoluble in water.
CCCCCC[N+]1=CN(C)C=C1
167.28
CHEBI:61340
98%
Videos
Safety and Handling

Safety and Handling

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: III

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

Product Identifier
  • 1-n-Hexyl-3-methylimidazolium chloride

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Supplemental information
  • MIXTURE LIST-Contains : 1-Hexyl-3-methylimidazolium chloride

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.