missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthalenemethanol, 98+%, Thermo Scientific Chemicals

Product Code. 11364574
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4780-79-4
Molecular Formula C11H10O
Molecular Weight (g/mol) 158.20
MDL Number MFCD00004044
InChI Key PBLNHHSDYFYZNC-UHFFFAOYSA-N
Synonym 1-naphthalenemethanol, 1-naphthylmethanol, 1-hydroxymethylnaphthalene, naphthalene-1-methanol, 1-naphthalene methanol, 1-menaphthyl alcohol, 1-naphthylmethyl alcohol, naphthalen-1-yl-methanol, naphthalen-1-yl methanol, 1-naphthylenemethanol
PubChem CID 20908
ChEBI CHEBI:38137
IUPAC Name naphthalen-1-ylmethanol
SMILES OCC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point 58°C to 63°C
Boiling Point 301°C (715 mmHg)
Quantity 50 g
Beilstein 2042532
Formula Weight 158.2
Percent Purity ≥98%
Chemical Name or Material 1-Naphthalenemethanol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'