Learn More
1-Naphthoyl chloride, 98%, Thermo Scientific Chemicals
Description
1-Naphthoyl chloride has been used as derivatization reagent in analysis of short-chained dodecyl alcohol ethoxylates and dodecyl alcohol by solid-phase extraction combined with dispersive liquid-liquid micro extraction method, as fluorescent labeling reagent in determination of T-2 and HT-2 toxins by HPLC with fluorescence detection, in Arndt-Eistert synthesis in the presence of trimethylsilyldiazomethane and in preparation of 2-ethyl-1-pentyl-3-(1-naphthoyl)indole.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
1-Naphthoyl chloride has been used as derivatization reagent in analysis of short-chained dodecyl alcohol ethoxylates and dodecyl alcohol by solid-phase extraction combined with dispersive liquid-liquid micro extraction method, as fluorescent labeling reagent in determination of T-2 and HT-2 toxins by HPLC with fluorescence detection, in Arndt-Eistert synthesis in the presence of trimethylsilyldiazomethane and in preparation of 2-ethyl-1-pentyl-3-(1-naphthoyl)indole.
Solubility
Reacts with water.
Notes
Moisture sensitive. Store away from oxidizing agents, bases, amines, alcohols, active metals and water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.

Specifications
Specifications
Melting Point | 16°C to 19°C |
Density | 1.265 |
Boiling Point | 118°C (0.1 mmHg) |
Flash Point | >110°C (230°F) |
Odor | Pungent |
Refractive Index | 1.6525 |
Quantity | 10 g |
UN Number | UN3265 |
Beilstein | 775785 |
Sensitivity | Moisture Sensitive |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0447081 | Certificate of Analysis | 30/08/2024 | A14460.18,A14460.09,A14460.30 |
A0446849 | Certificate of Analysis | 30/01/2023 | A14460.18,A14460.09,A14460.30 |
A0433454 | Certificate of Analysis | 30/01/2023 | A14460.18,A14460.09,A14460.30 |
10153088 | Certificate of Analysis | 22/11/2021 | A14460.18,A14460.09,A14460.30 |
10180709 | Certificate of Analysis | 22/11/2021 | A14460.18,A14460.09,A14460.30 |
Safety and Handling

- 1-Naphthoyl chloride
- Danger
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : 1-Naphthalenecarbonyl chloride
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.