missing translation for 'onlineSavingsMsg'
Learn More
1-Naphthyl phosphate disodium salt hydrate, 99%, Thermo Scientific Chemicals Product Code.: 11429917

1-Naphthyl phosphate disodium salt hydrate, 99%, Thermo Scientific Chemicals

Product Code. 11429917
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11429917

Brand: Thermo Scientific Alfa Aesar B22866.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Naphthyl phosphate disodium salt hydrate, 99%, C10H9Na2O5P, CAS Number-207569-06-0

1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207569-06-0
Molecular Formula C10H7Na2O4P
Molecular Weight (g/mol) 268.12
MDL Number MFCD00041007
InChI Key QYURIFWAOPAPAJ-UHFFFAOYSA-L
Synonym 1-naphthyl phosphate disodium salt hydrate, sodium naphthalen-1-yl phosphate hydrate, disodium 1-naphthyl phosphate hydrate, disodium hydrate naphthalen-1-yl phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h7o4p.2na.h2o, 1-naphthyl?phosphate?disodium?salt?hydrate, 1-naphthyl phosphate disodium salt monohydrate, dihydrogen naphthyl phosphate, oxamethane, sodium salt, sodium salt
PubChem CID 24212014
IUPAC Name disodium;naphthalen-1-yl phosphate;hydrate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 4895811
Solubility Information Soluble in water.
Formula Weight 268.12 (Anhydrous)
Percent Purity 99%
Chemical Name or Material 1-Naphthyl phosphate disodium salt hydrate

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10246047 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberB22866.03,B22866.14,B22866.06
Lot Number10191240 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22866.03,B22866.14,B22866.06
Lot Number10200888 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22866.03,B22866.14,B22866.06
Lot Number10167433 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22866.03,B22866.14,B22866.06
Lot Number10115481 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB22866.03,B22866.14,B22866.06
compliance-icons
Product Identifier
  • 1-Naphthyl phosphate disodium salt hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : 1-Naphthylphosphatedisodium salt hydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Naphthyl phosphate disodium salt hydrate, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.