Learn More
1-Naphthyl phosphate, monosodium salt monohydrate, 98+%, Thermo Scientific Chemicals
£23.90 - £897.00
Chemical Identifiers
CAS | 81012-89-7 |
---|---|
Molecular Formula | C10H7O4P |
Molecular Weight (g/mol) | 222.14 |
MDL Number | MFCD00150615 |
InChI Key | YNXICDMQCQPQEW-UHFFFAOYSA-L |
Synonym | 1-naphthyl phosphate monosodium salt monohydrate, sodium naphthalen-1-yl hydrogenphosphate hydrate, alpha-naphthyl acid phosphate monosodium, sodium hydrate naphthalen-1-yl hydrogen phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h8po4.na.h2o, sodium 1-naphthyl phosphate monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, 1-naphthyl phosphate monosodium salt monohydrate hplc |
PubChem CID | 45055387 |
SMILES | [O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10434345
|
Thermo Scientific Acros
226890010 |
1 g | Glass bottle |
£23.90
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10124760
|
Thermo Scientific Acros
226890050 |
5 g | Glass bottle |
£72.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10752685
|
Thermo Scientific Acros
226890250 |
25 g | Glass bottle |
£244.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15169253
|
Thermo Scientific Acros
226891000 |
100 g | Glass bottle |
£897.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
81012-89-7 | |
222.14 | |
YNXICDMQCQPQEW-UHFFFAOYSA-L | |
45055387 |
C10H7O4P | |
MFCD00150615 | |
1-naphthyl phosphate monosodium salt monohydrate, sodium naphthalen-1-yl hydrogenphosphate hydrate, alpha-naphthyl acid phosphate monosodium, sodium hydrate naphthalen-1-yl hydrogen phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h8po4.na.h2o, sodium 1-naphthyl phosphate monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, 1-naphthyl phosphate monosodium salt monohydrate hplc | |
[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1 |
Specifications
81012-89-7 | |
100.0 | |
White | |
Authentic | |
C10H7O4P | |
MFCD00150615 | |
(5% in water) Clear colorless to light yellow | |
[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1 | |
222.14 | |
264.15 | |
Fine Powder |
98.0 | |
189.0°C to 191.0°C | |
3.5 to 4.5 (5% aq. soln.) | |
98+% | |
C10H7OP(O)(OH)ONa·H2O | |
1-naphthyl phosphate monosodium salt monohydrate, sodium naphthalen-1-yl hydrogenphosphate hydrate, alpha-naphthyl acid phosphate monosodium, sodium hydrate naphthalen-1-yl hydrogen phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h8po4.na.h2o, sodium 1-naphthyl phosphate monohydrate, 1-naphthyl phosphate, monosodium salt monohydrate, naphthalen-1-yloxy sodiooxy phosphinic acid hydrate, 1-naphthyl phosphate monosodium salt monohydrate hplc | |
YNXICDMQCQPQEW-UHFFFAOYSA-L | |
naphthalen-1-yl dihydrogen phosphate;sodium;hydrate | |
45055387 | |
98+% | |
1-Naphthyl phosphate, monosodium salt monohydrate, 98+% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get med
GHS Signal Word: Warning
missing translation for 'tsca' : TSCA
- 1-Naphthyl phosphate, monosodium salt, monohydrate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only