missing translation for 'onlineSavingsMsg'
Learn More

1-Naphthyl trifluoromethanesulfonate, 97%, Thermo Scientific Chemicals

POA - £152.00

Chemical Identifiers

CAS 99747-74-7
Molecular Formula C11H7F3O3S
Molecular Weight (g/mol) 276.229
MDL Number MFCD00192340
InChI Key WQWUQDVFRYMMCY-UHFFFAOYSA-N
Synonym 1-naphthyl trifluoromethanesulfonate, 1-naphthyl triflate, 1-naphthyl-triflate, 1-naphthalenyl triflate, acmc-20aot0, 1-trifluoromethanesulfonyl naphthalene
PubChem CID 378884
IUPAC Name naphthalen-1-yl trifluoromethanesulfonate
SMILES C1=CC=C2C(=C1)C=CC=C2OS(=O)(=O)C(F)(F)F
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15431238
View Documents
Thermo Scientific Alfa Aesar
H55091.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15411238
View Documents
Thermo Scientific Alfa Aesar
H55091.03
1 g
£43.60
1g
Please to purchase this item. Need a web account? Register with us today!
15421238
View Documents
Thermo Scientific Alfa Aesar
H55091.06
5 g
£152.00
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

99747-74-7
276.229
WQWUQDVFRYMMCY-UHFFFAOYSA-N
378884
C1=CC=C2C(=C1)C=CC=C2OS(=O)(=O)C(F)(F)F
C11H7F3O3S
MFCD00192340
1-naphthyl trifluoromethanesulfonate, 1-naphthyl triflate, 1-naphthyl-triflate, 1-naphthalenyl triflate, acmc-20aot0, 1-trifluoromethanesulfonyl naphthalene
naphthalen-1-yl trifluoromethanesulfonate

Specifications

99747-74-7
1.405 g/mL
113°C (235°F)
1.52
UN3265
WQWUQDVFRYMMCY-UHFFFAOYSA-N
naphthalen-1-yl trifluoromethanesulfonate
378884
97%
1-Naphthyl trifluoromethanesulfonate
Red
97°C to 98°C (0.3 mmHg)
C11H7F3O3S
MFCD00192340
1-naphthyl trifluoromethanesulfonate, 1-naphthyl triflate, 1-naphthyl-triflate, 1-naphthalenyl triflate, acmc-20aot0, 1-trifluoromethanesulfonyl naphthalene
C1=CC=C2C(=C1)C=CC=C2OS(=O)(=O)C(F)(F)F
276.229
276.23
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.