Learn More
1-Naphthylacetonitrile 97%, Thermo Scientific™
£22.80
Chemical Identifiers
CAS | 132-75-2 |
---|---|
Molecular Formula | C12H9N |
Molecular Weight (g/mol) | 167.211 |
MDL Number | MFCD00004041 |
InChI Key | OQRMWUNUKVUHQO-UHFFFAOYSA-N |
Synonym | 1-naphthylacetonitrile, 2-naphthalen-1-yl acetonitrile, 1-naphthyl acetonitrile, 1-naphthaleneacetonitrile, acetonitrile, 1-naphthyl, alpha-naphthylacetonitrile, alpha-naphthyl acetonitrile, 1-naphthylmethylcyanide, .alpha.-naphthylacetonitrile, 2-1-naphthyl acetonitrile |
PubChem CID | 8596 |
IUPAC Name | 2-naphthalen-1-ylacetonitrile |
SMILES | C1=CC=C2C(=C1)C=CC=C2CC#N |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10011300
|
Thermo Scientific Acros
128230250 |
25 g | Plastic Bottle | ||||||
Description
Chemical Identifiers
132-75-2 | |
167.211 | |
OQRMWUNUKVUHQO-UHFFFAOYSA-N | |
8596 | |
C1=CC=C2C(=C1)C=CC=C2CC#N |
C12H9N | |
MFCD00004041 | |
1-naphthylacetonitrile, 2-naphthalen-1-yl acetonitrile, 1-naphthyl acetonitrile, 1-naphthaleneacetonitrile, acetonitrile, 1-naphthyl, alpha-naphthylacetonitrile, alpha-naphthyl acetonitrile, 1-naphthylmethylcyanide, .alpha.-naphthylacetonitrile, 2-1-naphthyl acetonitrile | |
2-naphthalen-1-ylacetonitrile |
Specifications
132-75-2 | |
100.0 | |
Yellow to Brown | |
>112°C | |
97% | |
C10H7CH2CN | |
MFCD00004041 | |
1-naphthylacetonitrile, 2-naphthalen-1-yl acetonitrile, 1-naphthyl acetonitrile, 1-naphthaleneacetonitrile, acetonitrile, 1-naphthyl, alpha-naphthylacetonitrile, alpha-naphthyl acetonitrile, 1-naphthylmethylcyanide, .alpha.-naphthylacetonitrile, 2-1-naphthyl acetonitrile | |
C1=CC=C2C(=C1)C=CC=C2CC#N | |
167.211 | |
167.21 | |
Liquid After Melting |
96.0 | |
33.0°C to 35.0°C | |
191.0°C to 194.0°C (18.0 mmHg) | |
Authentic | |
C12H9N | |
1.6182 to 1.6202 | |
09, 667 | |
OQRMWUNUKVUHQO-UHFFFAOYSA-N | |
2-naphthalen-1-ylacetonitrile | |
8596 | |
97% | |
1-Naphthylacetonitrile, 97% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
Harmful in contact with skin.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 205-078-1
missing translation for 'rtecsNumber' : AM1240000
missing translation for 'tsca' : TSCA
- 1-Naphthylacetonitrile
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only