missing translation for 'onlineSavingsMsg'
Learn More

1-Nitropropane, 98%, Thermo Scientific Chemicals

Catalog Number 10396720
Click to view available options
Quantity:
5 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
108-03-2
C3H7NO2
89.09
MFCD00007407
JSZOAYXJRCEYSX-UHFFFAOYSA-N
propane, 1-nitro, nitropropane, propane, nitro, ccris 1329, dsstox_cid_980, dsstox_rid_75902, dsstox_gsid_20980, nitro-propane, n-nitropropane, 3-nitropropane
7903
CHEBI:76261
1-nitropropane
CCC[N+](=O)[O-]
This item is not returnable. View return policy
108-03-2
C3H7NO2
89.09
MFCD00007407
JSZOAYXJRCEYSX-UHFFFAOYSA-N
propane, 1-nitro, nitropropane, propane, nitro, ccris 1329, dsstox_cid_980, dsstox_rid_75902, dsstox_gsid_20980, nitro-propane, n-nitropropane, 3-nitropropane
7903
CHEBI:76261
1-nitropropane
CCC[N+](=O)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point -108°C
Density 0.9900g/mL
Boiling Point 132°C
Flash Point 33°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2NO2
Refractive Index 1.4005 to 1.4025
Quantity 5 g
Beilstein 01, 115
Fieser 01,745
Merck Index 15, 6714
Specific Gravity 0.99
Solubility Information Solubility in water: 1.4mL/100mL (20°C). Other solubilities: miscible with many organic solvents
Viscosity 0.844 mPa.s (20°C)
Formula Weight 89.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Nitropropane, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'