missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Nonylamine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
500 g
Description
1-Nonylamine is used as pharmaceutical intermediate. It is also used as a building block.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Nonylamine is used as pharmaceutical intermediate. It is also used as a building block.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
1-Nonylamine is used as pharmaceutical intermediate. It is also used as a building block.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents and carbon dioxide.
Specifications
Specifications
Melting Point | -1°C |
Density | 0.784 |
Boiling Point | 200°C to 202°C |
Flash Point | 62°C (143°F) |
Odor | Amine-like |
Refractive Index | 1.433 |
Quantity | 25 g |
UN Number | UN2735 |
Beilstein | 1733633 |
Sensitivity | Air Sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'