missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecanesulfonic acid, sodium salt, 90%, Thermo Scientific Chemicals

Product Code. 10698234
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13893-34-0
Molecular Formula C18H37NaO3S
Molecular Weight (g/mol) 356.54
MDL Number MFCD00051526
InChI Key KBAFDSIZQYCDPK-UHFFFAOYSA-M
Synonym sodium 1-octadecanesulfonate, sodium octadecane-1-sulfonate, sodium stearylsulfonate, sodium octadecanesulfonate, 1-octadecanesulfonic acid sodium salt, sodium octadecane-1-sulphonate, 1-octadecanesulfonic acid, sodium salt, sodium sterylsulfonate, sodium oxadecanesulfonate, sodium n-octadecylsulfonate
PubChem CID 23668003
IUPAC Name sodium;octadecane-1-sulfonate
SMILES CCCCCCCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]

Specifications

CAS Min % 88.0
CAS Max % 100.0
Melting Point 163°C to 165°C
Color White
Elemental Analysis 88% min. (C, H)
Loss on Drying 5% max. (1 G, 105°C, 1 Hrs, 760mm)
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula CH3(CH2)17SO3Na
Quantity 1 g
Solubility Information Solubility in water: soluble
Formula Weight 356.54
Percent Purity 90%
Physical Form Powder
Chemical Name or Material 1-Octadecanesulfonic acid, sodium salt
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'