missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecene, 90%, tech.

Product Code. 11984421 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
Unit Size:
1 L
2.5 L
This item is not returnable. View return policy

Product Code. 11984421

Brand: Thermo Scientific Acros 129310025

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 112-88-9 | C18H36 | 252.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-88-9
Molecular Formula C18H36
Molecular Weight (g/mol) 252.48
InChI Key CCCMONHAUSKTEQ-UHFFFAOYSA-N
Synonym 1-octadecene, octadecene, alpha-octadecene, .alpha.-octadecene, unii-h5zuq6v4ak, octadecene-1, h5zuq6v4ak, alkenes, c14-20 .alpha., 1-octadecene, analytical standard, linear octadecene
PubChem CID 8217
ChEBI CHEBI:30824
IUPAC Name octadec-1-ene
SMILES CCCCCCCCCCCCCCCCC=C

Specifications

Melting Point 17.5°C
Color Colorless to Amber
Density 0.7890g/mL
Boiling Point 315°C
Flash Point 154°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula CH3(CH2)15CH=CH2
Refractive Index 1.4435 to 1.4455
Quantity 2.5 L
Beilstein 01,226
Specific Gravity 0.789
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether, petroleum and, coal-tar solvents
Viscosity 3.6 mm2/s (40°C)
Formula Weight 252.48
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 1-Octadecene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Octadecene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.