missing translation for 'onlineSavingsMsg'
Learn More

1-Octadecene, 90%, tech., Thermo Scientific Chemicals

Catalog Number p-4509848
31.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass bottle
112-88-9
C18H36
252.48
CCCMONHAUSKTEQ-UHFFFAOYSA-N
1-octadecene, octadecene, alpha-octadecene, .alpha.-octadecene, unii-h5zuq6v4ak, octadecene-1, h5zuq6v4ak, alkenes, c14-20 .alpha., 1-octadecene, analytical standard, linear octadecene
8217
CHEBI:30824
octadec-1-ene
CCCCCCCCCCCCCCCCC=C
112-88-9
C18H36
252.48
CCCMONHAUSKTEQ-UHFFFAOYSA-N
1-octadecene, octadecene, alpha-octadecene, .alpha.-octadecene, unii-h5zuq6v4ak, octadecene-1, h5zuq6v4ak, alkenes, c14-20 .alpha., 1-octadecene, analytical standard, linear octadecene
8217
CHEBI:30824
octadec-1-ene
CCCCCCCCCCCCCCCCC=C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 17.5°C
Color Colorless to Amber
Density 0.7890g/mL
Boiling Point 315°C
Flash Point 154°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Linear Formula CH3(CH2)15CH=CH2
Refractive Index 1.4435 to 1.4455
Quantity 1 L
Beilstein 01,226
Specific Gravity 0.789
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone, ether, petroleum and, coal-tar solvents
Viscosity 3.6 mm2/s (40°C)
Formula Weight 252.48
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 1-Octadecene
Show More Show Less
compliance-icons
Product Identifier
  • 1-Octadecene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'