missing translation for 'onlineSavingsMsg'
Learn More

1-Octanesulfonic acid sodium salt, monohydrate, 99.7%, For HPLC analysis, MP Biomedicals™

Brand:  MP Biomedicals 0215177880

Code : NEW

Additional Details : CAS Number : 5324-84-5 Weight : 0.10000kg

 View more versions of this product

Product Code. 11432572

  • £339.20 / 100g
Estimated Shipment: 10-10-2024
to see stock.

Description

Description

Specifications

Specifications

1-Octanesulfonic Acid, Sodium Salt, Monohydrate
Crystalline Powder
White
Store at room temperature (15°C to 30°C)
100 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers