missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Octanol, ≥99%, ACS Spectrophotometric Grade, Honeywell Riedel-de Haën™
Click to view available options
Quantity:
1 L
2 L
Packaging:
Glass bottle
Description
- ACS grade quality reagents and solvents tested according to the specific use for spectrophotometric applications.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -15°C |
Density | 0.827g/mL at 25°C |
Boiling Point | 194°C |
Flash Point | 80°C (176°F) |
Packaging | Glass bottle |
Linear Formula | CH3(CH2)7OH |
Refractive Index | 1.429 |
Quantity | 1 L |
Beilstein | 1697461 |
Vapor Density | 4.5 (vs air) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'