missing translation for 'onlineSavingsMsg'
Learn More

1-Octyl isothiocyanate, 97%

Product Code. 11421848
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11421848

Brand: Thermo Scientific Alfa Aesar A14501.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4430-45-9 | C9H17NS | 171.30 g/mol

1-Octyl isothiocyanate can be used to produce 1-(6-methyl-quinolin-2-yl)-3-octyl-thiourea. It is used as intermediates in pharmaceutical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octyl isothiocyanate can be used to produce 1-(6-methyl-quinolin-2-yl)-3-octyl-thiourea. It is used as intermediates in pharmaceutical industry.

Solubility
It hydrolyzes in water.

Notes
It is moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4430-45-9
Molecular Formula C9H17NS
Molecular Weight (g/mol) 171.30
MDL Number MFCD00041139
InChI Key YEZHGQZHWKJPCM-UHFFFAOYSA-N
Synonym octyl isothiocyanate, n-octyl isothiocyanate, octylisothiocyanate, 1-octyl isothiocyanate, octanisothiocyanate, 1-isothiocyanatooctan, octane, 1-isothiocyanato, isothiocyanic acid, octyl ester, n-octylisothiocyanate, acmc-1ajcy
PubChem CID 78161
IUPAC Name 1-isothiocyanatooctane
SMILES CCCCCCCCN=C=S

Specifications

Density 0.88
Boiling Point 124°C to 126°C (7 mmHg)
Flash Point 69°C (156°F)
Odor Pungent
Refractive Index 1.49
Quantity 5 g
UN Number UN2922
Beilstein 1760783
Sensitivity Moisture sensitive
Solubility Information It hydrolyzes in water.
Formula Weight 171.31
Percent Purity 97%
Chemical Name or Material 1-Octyl isothiocyanate
Show More Show Less
compliance-icons
Product Identifier
  • 1-Octyl isothiocyanate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Octyl isothiocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.