missing translation for 'onlineSavingsMsg'
Learn More

1-Octylphosphonic acid, 99%

Product Code. 11339425
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11339425

missing translation for 'mfr': Thermo Scientific Alfa Aesar L18586.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4724-48-5 | C8H19O3P | 194.211 g/mol

1-Octylphosphonic acid is a basic building block and used in the preparation of various pharmaceutical compounds and chemical products. It is supported with calcium hydroxyapatite and used in the dental field as corrosion inhibitor and organic building block. It acts as a corrosion inhibitor in metallic household surfaces such as faucets and drains as well as prevents the rusting of can or dispenser. It is also used in the synthesis of octyl-phosphonic acid dimethyl ester by reacting with iodomethane in presence of CS2 CO3.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Octylphosphonic acid is a basic building block and used in the preparation of various pharmaceutical compounds and chemical products. It is supported with calcium hydroxyapatite and used in the dental field as corrosion inhibitor and organic building block. It acts as a corrosion inhibitor in metallic household surfaces such as faucets and drains as well as prevents the rusting of can or dispenser. It is also used in the synthesis of octyl-phosphonic acid dimethyl ester by reacting with iodomethane in presence of CS2 CO3.

Solubility
Slightly soluble in organic solvents and water.

Notes
Incompatible with strong oxidizing agents. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4724-48-5
Molecular Formula C8H19O3P
Molecular Weight (g/mol) 194.211
MDL Number MFCD00015841
InChI Key NJGCRMAPOWGWMW-UHFFFAOYSA-N
Synonym n-octylphosphonic acid, 1-octylphosphonic acid, phosphonic acid, octyl, phosphonic acid, p-octyl, unii-95c2aw87bo, octylphosphonicacid, acmc-209uni, p-octyl-phosphonic acid, octylphosphonic acid, dsstox_cid_24679
PubChem CID 78452
IUPAC Name octylphosphonic acid
SMILES CCCCCCCCP(=O)(O)O

Specifications

Melting Point 100°C to 102°C
Quantity 1 g
Beilstein 1764242
Solubility Information Slightly soluble in organic solvents and water.
Formula Weight 194.21
Percent Purity 99%
Chemical Name or Material 1-Octylphosphonic acid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1-Octylphosphonic acid
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H373-May cause damage to organs.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1-Octylphosphonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.