missing translation for 'onlineSavingsMsg'
Learn More

1-(p-Toluenesulfonyl)imidazole, 98+%

Product Code. 15490538
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 15490538

Brand: Thermo Scientific Alfa Aesar H54191.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2232-08-8 | C10H10N2O2S | 222.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2232-08-8
Molecular Formula C10H10N2O2S
Molecular Weight (g/mol) 222.26
MDL Number MFCD00005285
InChI Key YJYMYJRAQYREBT-UHFFFAOYSA-N
Synonym 1-p-toluenesulfonyl imidazole, 1-tosyl-1h-imidazole, n-tosylimidazole, 1-4-methylphenyl sulfonyl-1h-imidazole, 1-tosylimidazole, 1-p-toluenesulphonyl imidazole, 1-p-tolylsulfonyl imidazole, 1h-imidazole, 1-4-methylphenyl sulfonyl, 1-toluene-p-sulphonyl imidazole, 1-4-methylbenzenesulfonyl imidazole
PubChem CID 75219
IUPAC Name 1-(4-methylphenyl)sulfonylimidazole
SMILES CC1=CC=C(C=C1)S(=O)(=O)N1C=CN=C1

Specifications

Melting Point 76°C to 78°C
Quantity 5 g
Beilstein 612451
Sensitivity Moisture sensitive
Formula Weight 222.26
Percent Purity ≥98%
Chemical Name or Material 1-(p-Toluenesulfonyl)imidazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.