missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanesulfonic acid, sodium salt, 98+%

Product Code. 10698624 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10698624

Brand: Thermo Scientific Acros 206111000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22767-49-3 | C5H11NaO3S | 174.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22767-49-3
Molecular Formula C5H11NaO3S
Molecular Weight (g/mol) 174.19
MDL Number MFCD00007541,MFCD00149548
InChI Key ROBLTDOHDSGGDT-UHFFFAOYSA-M
Synonym sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt
PubChem CID 23664617
IUPAC Name sodium;pentane-1-sulfonate
SMILES [Na+].CCCCCS([O-])(=O)=O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point >300.0°C
Color White to Yellow
Loss on Drying 2 % max. (1 g, 120°C)
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Linear Formula CH3(CH2)4SO3Na
Quantity 100 g
Beilstein 04, III, 23
Solubility Information (10% in water at 25°C) Clear colorless
Formula Weight 174.2
Percent Purity 98+%
Physical Form Crystals
Chemical Name or Material 1-Pentanesulfonic acid, sodium salt
Show More Show Less
compliance-icons
Product Identifier
  • 1-Pentanesulfonic acid, sodium salt
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.