Learn More
1-Pentanesulfonic acid, sodium salt, 99+%, Ion pair chromatography, anhydrous, Thermo Scientific Chemicals
£119.00 - £1551.00
Chemical Identifiers
CAS | 22767-49-3 |
---|---|
Molecular Formula | C5H11NaO3S |
Molecular Weight (g/mol) | 174.19 |
MDL Number | MFCD00007541,MFCD00149548 |
InChI Key | ROBLTDOHDSGGDT-UHFFFAOYSA-M |
Synonym | sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt |
PubChem CID | 23664617 |
IUPAC Name | sodium;pentane-1-sulfonate |
SMILES | [Na+].CCCCCS([O-])(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10024523
|
Thermo Scientific Acros
396650250 |
25 g | Glass bottle |
£119.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10116033
|
Thermo Scientific Acros
396651000 |
100 g | Glass bottle |
£385.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11914571
|
Thermo Scientific Acros
396655000 |
500 g | Glass Bottle |
£1551.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ion pair chromatographyChemical Identifiers
22767-49-3 | |
174.19 | |
ROBLTDOHDSGGDT-UHFFFAOYSA-M | |
23664617 | |
[Na+].CCCCCS([O-])(=O)=O |
C5H11NaO3S | |
MFCD00007541,MFCD00149548 | |
sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt | |
sodium;pentane-1-sulfonate |
Specifications
22767-49-3 | |
>300.0°C | |
2% (150°C) max. | |
99+% | |
CH3(CH2)4SO3Na | |
sodium 1-pentanesulfonate, sodium pentane-1-sulfonate, sodium pentanesulfonate, 1-pentanesulfonic acid sodium salt, 1-pentanesulfonic acid, sodium salt, 1-pentylsodiumsulfonate, ipc-alks-5, 1-pentanesulfonate, sodium, pentanesulfonic acid na-salt, pentanesulfonic acidsodiumsalt | |
ROBLTDOHDSGGDT-UHFFFAOYSA-M | |
sodium;pentane-1-sulfonate | |
23664617 | |
99+% | |
Crystalline Powder |
0.02 max. (C=0.5 M in water) at 260nm, 0.04 max. (C=0.5 M in water) at 230nm, 0.06 max. C=0.5 M in water at 220nm, 0.1 max. (C=0.5 M in water) at 210nm | |
White | |
Authentic | |
C5H11NaO3S | |
MFCD00007541,MFCD00149548 | |
Solubility in water: soluble. | |
[Na+].CCCCCS([O-])(=O)=O | |
174.19 | |
174.2 | |
Ion Pair Chromatography | |
1-Pentanesulfonic acid, sodium salt |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 245-208-4
- 1-Pentanesulfonic acid, sodium salt
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only