missing translation for 'onlineSavingsMsg'
Learn More

1-Pentanol, 99%, pure, Thermo Scientific Chemicals

Product Code. 10013313
Click to view available options
Quantity:
25 mL
1 L
2.5 L
25 L
Packaging:
Glass Bottle
Metal drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-41-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
Synonym 1-pentanol, amyl alcohol, n-amyl alcohol, n-pentanol, pentanol, pentyl alcohol, butylcarbinol, 1-pentyl alcohol, amylol, n-butylcarbinol
PubChem CID 6276
ChEBI CHEBI:44884
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -78.0°C
Density 0.8110g/mL
Boiling Point 137.0°C to 139.0°C
Flash Point 49°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Metal drum
Linear Formula CH3(CH2)4OH
Refractive Index 1.4083 to 1.4103
Quantity 25 L
Beilstein 01, 383
Merck Index 15, 7230
Specific Gravity 0.811
Solubility Information Solubility in water: 22g/L (22°C). Other solubilities: miscible with alcohol, benzene and ether, soluble in most organic solvents
Viscosity 4 mPa.s (20°C)
Formula Weight 88.15
Percent Purity 99%
Grade Pure
Physical Form Liquid
Chemical Name or Material 1-Pentanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Pentanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'