missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Pentanol, ReagentPlus™, ≥99%, Honeywell™
Click to view available options
Quantity:
100 mL
500 mL
1 L
2.5 L
4 L
20 L
Packaging:
Aluminum Bottle
Glass Bottle
Stainless Steel Can
Description
- ≥95% for general laboratory use.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | -78°C |
Color | Colorless |
Density | 0.811g/mL at 25°C |
Boiling Point | 136°C to 138°C |
Flash Point | 49°C (120.2°F) |
Packaging | Glass Bottle |
Linear Formula | CH3(CH2)4OH |
Refractive Index | n20/D 1.409(lit.) |
Quantity | 100 mL |
UN Number | UN1105 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'