missing translation for 'onlineSavingsMsg'
Learn More

(1-Pentyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Product Code. 11464204
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates.

Solubility
Partly miscible in water. Soluble in methanol and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21406-61-1
Molecular Formula C23H26BrP
Molecular Weight (g/mol) 413.34
MDL Number MFCD00031630
InChI Key VAUKWMSXUKODHR-UHFFFAOYSA-M
Synonym pentyltriphenylphosphonium bromide, amyltriphenylphosphonium bromide, n-amyl triphenylphosphonium bromide, pentyltriphenylphosphanium bromide, 1-pentyl triphenylphosphonium bromide, triphenyl-n-amylphosphonium bromide, pentyl triphenyl phosphanium bromide, phosphonium, pentyltriphenyl-, bromide, pentyl triphenyl phosphonium bromide, pubchem15722
PubChem CID 2724565
IUPAC Name pentyl(triphenyl)phosphanium;bromide
SMILES [Br-].CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 164°C to 166°C
Quantity 25 g
Beilstein 3580453
Sensitivity Hygroscopic
Solubility Information Partly miscible in water. Soluble in methanol and chloroform.
Formula Weight 413.33
Percent Purity 98%
Chemical Name or Material (1-Pentyl)triphenylphosphonium bromide
compliance-icons
Product Identifier
  • (1-Pentyl)triphenylphosphonium bromide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Pentyltriphenylphosphonium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.