missing translation for 'onlineSavingsMsg'
Learn More

(1-Pentyl)triphenylphosphonium bromide, 98%, Thermo Scientific Chemicals

Product Code. 11484204
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediates.

Solubility
Partly miscible in water. Soluble in methanol and chloroform.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21406-61-1
Molecular Formula C23H26BrP
Molecular Weight (g/mol) 413.34
MDL Number MFCD00031630
InChI Key VAUKWMSXUKODHR-UHFFFAOYSA-M
Synonym pentyltriphenylphosphonium bromide, amyltriphenylphosphonium bromide, n-amyl triphenylphosphonium bromide, pentyltriphenylphosphanium bromide, 1-pentyl triphenylphosphonium bromide, triphenyl-n-amylphosphonium bromide, pentyl triphenyl phosphanium bromide, phosphonium, pentyltriphenyl-, bromide, pentyl triphenyl phosphonium bromide, pubchem15722
PubChem CID 2724565
IUPAC Name pentyl(triphenyl)phosphanium;bromide
SMILES [Br-].CCCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 164°C to 166°C
Quantity 500 g
Beilstein 3580453
Sensitivity Hygroscopic
Solubility Information Partly miscible in water. Soluble in methanol and chloroform.
Formula Weight 413.33
Percent Purity 98%
Chemical Name or Material (1-Pentyl)triphenylphosphonium bromide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'