missing translation for 'onlineSavingsMsg'
Learn More
1-Phenyl-1,2-ethanediol, 97%, Thermo Scientific Chemicals Product Code.: 10506941

1-Phenyl-1,2-ethanediol, 97%, Thermo Scientific Chemicals

Product Code. 10506941
5 g, Glass bottle
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10506941

Brand: Thermo Scientific Acros 130590050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-56-1
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00003546
InChI Key PWMWNFMRSKOCEY-UHFFFAOYSA-N
Synonym 1-phenyl-1,2-ethanediol, styrene glycol, phenylethylene glycol, phenylethanediol, phenyl glycol, 1,2-ethanediol, 1-phenyl, phenyl-1,2-ethanediol, styrolyl alcohol, fenylglycol, 1-phenylethylene glycol
PubChem CID 7149
IUPAC Name 1-phenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(CO)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 61°C to 67°C
Color White to Beige
Boiling Point 272°C to 274°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2CH(C6H5)OH
Quantity 5 g
Beilstein 06, 907
Merck Index 15, 8991
Formula Weight 138.17
Percent Purity 97%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 1-Phenyl-1, 2-ethanediol
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0410954 Certificate TypeCertificate of Analysis Date04/02/2022 Catalog Number
Lot NumberA0401792 Certificate TypeCertificate of Analysis Date28/04/2021 Catalog Number
Product Identifier
  • 1-Phenyl-1,2-ethanediol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Phenyl-1,2-ethanediol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.