missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-1,2-propanedione-2-oxime, 99%, Thermo Scientific Chemicals
£29.50 - £55.10
Chemical Identifiers
CAS | 119-51-7 |
---|---|
Molecular Formula | C9H9NO2 |
Molecular Weight (g/mol) | 163.18 |
MDL Number | MFCD00002115 |
InChI Key | YPINLRNGSGGJJT-JXMROGBWSA-N |
Synonym | 1-phenyl-1,2-propanedione-2-oxime, 2-hydroxyimino propiophenone, isonitrosopropiophenone, 2-hydroxyiminopropiophenone, 2-hydroxyimino-1-phenylpropan-1-one, propiophenone, isonitroso, 2-isonitrosopropiophenone, alpha-oximinopropiophenone, 1-phenyl-1,2-propanedione 2-oxime, .alpha.-oximinopropiophenone |
PubChem CID | 9566063 |
IUPAC Name | (2E)-2-hydroxyimino-1-phenylpropan-1-one |
SMILES | C\C(=N/O)C(=O)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10726803
|
Thermo Scientific Acros
205020100 |
10 g | Glass bottle |
£29.50
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10551841
|
Thermo Scientific Acros
205020250 |
25 g | Glass bottle |
£55.10
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
119-51-7 | |
163.18 | |
YPINLRNGSGGJJT-JXMROGBWSA-N | |
9566063 | |
C\C(=N/O)C(=O)C1=CC=CC=C1 |
C9H9NO2 | |
MFCD00002115 | |
1-phenyl-1,2-propanedione-2-oxime, 2-hydroxyimino propiophenone, isonitrosopropiophenone, 2-hydroxyiminopropiophenone, 2-hydroxyimino-1-phenylpropan-1-one, propiophenone, isonitroso, 2-isonitrosopropiophenone, alpha-oximinopropiophenone, 1-phenyl-1,2-propanedione 2-oxime, .alpha.-oximinopropiophenone | |
(2E)-2-hydroxyimino-1-phenylpropan-1-one |
Specifications
119-51-7 | |
100.0 | |
White to Yellow | |
99% | |
C6H5COC(=NOH)CH3 | |
07, III, 3464 | |
Solubility in water: 1.3g/L (20°C) | |
C\C(=N/O)C(=O)C1=CC=CC=C1 | |
163.18 | |
163.18 | |
Crystals |
98.5 | |
111.0°C to 115.0°C | |
Authentic | |
C9H9NO2 | |
MFCD00002115 | |
1-phenyl-1,2-propanedione-2-oxime, 2-hydroxyimino propiophenone, isonitrosopropiophenone, 2-hydroxyiminopropiophenone, 2-hydroxyimino-1-phenylpropan-1-one, propiophenone, isonitroso, 2-isonitrosopropiophenone, alpha-oximinopropiophenone, 1-phenyl-1,2-propanedione 2-oxime, .alpha.-oximinopropiophenone | |
YPINLRNGSGGJJT-JXMROGBWSA-N | |
(2E)-2-hydroxyimino-1-phenylpropan-1-one | |
9566063 | |
99% | |
1-Phenyl-1, 2-propanedione-2-oxime, 99% |
Safety and Handling
missing translation for 'einecsNumber' : 204-329-2
missing translation for 'rtecsNumber' : UH2975000
missing translation for 'tsca' : TSCA
RUO – Research Use Only