missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-1-cyclohexene, 95%, Thermo Scientific Chemicals
£22.55 - £71.30
Chemical Identifiers
CAS | 771-98-2 |
---|---|
Molecular Formula | C12H14 |
Molecular Weight (g/mol) | 158.24 |
MDL Number | MFCD00001542 |
InChI Key | WCMSFBRREKZZFL-UHFFFAOYSA-N |
Synonym | 1-phenyl-1-cyclohexene, 1-phenylcyclohexene, 1-phenylcyclohex-1-ene, benzene, 1-cyclohexen-1-yl, phenylcyclohexene, cyclohexenylbenzene, benzene, cyclohexenyl, 2,3,4,5-tetrahydro-1,1'-biphenyl, cyclohexene, 1-phenyl, trans-1-phenylcyclohexene |
PubChem CID | 13043 |
IUPAC Name | cyclohexen-1-ylbenzene |
SMILES | C1CCC(=CC1)C2=CC=CC=C2 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
771-98-2 | |
158.24 | |
WCMSFBRREKZZFL-UHFFFAOYSA-N | |
13043 | |
C1CCC(=CC1)C2=CC=CC=C2 |
C12H14 | |
MFCD00001542 | |
1-phenyl-1-cyclohexene, 1-phenylcyclohexene, 1-phenylcyclohex-1-ene, benzene, 1-cyclohexen-1-yl, phenylcyclohexene, cyclohexenylbenzene, benzene, cyclohexenyl, 2,3,4,5-tetrahydro-1,1'-biphenyl, cyclohexene, 1-phenyl, trans-1-phenylcyclohexene | |
cyclohexen-1-ylbenzene |
Specifications
771-98-2 | |
100.0 | |
Colorless to Yellow | |
251°C to 253°C | |
Authentic | |
C12H14 | |
1.5680 to 1.57 | |
05, II, 419 | |
1-phenyl-1-cyclohexene, 1-phenylcyclohexene, 1-phenylcyclohex-1-ene, benzene, 1-cyclohexen-1-yl, phenylcyclohexene, cyclohexenylbenzene, benzene, cyclohexenyl, 2,3,4,5-tetrahydro-1,1'-biphenyl, cyclohexene, 1-phenyl, trans-1-phenylcyclohexene | |
C1CCC(=CC1)C2=CC=CC=C2 | |
158.24 | |
158.24 | |
Liquid |
94.0 | |
-11°C | |
0.9900g/mL | |
103°C | |
94% min. (GC) | |
C6H5C6H9 | |
MFCD00001542 | |
0.99 | |
WCMSFBRREKZZFL-UHFFFAOYSA-N | |
cyclohexen-1-ylbenzene | |
13043 | |
95% | |
1-Phenyl-1-cyclohexene, 95% |
Safety and Handling
missing translation for 'einecsNumber' : 212-242-6
missing translation for 'rtecsNumber' : GW6600000
missing translation for 'tsca' : TSCA
RUO – Research Use Only