missing translation for 'onlineSavingsMsg'
Learn More

1-Phenyl-1-pyridin-2-ylmethanamine dihydrochloride, 95%, Thermo Scientific™

Product Code. 10252392
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 59575-91-6
Molecular Formula C12H13ClN2
Molecular Weight (g/mol) 220.70
MDL Number MFCD00102147
InChI Key GVGSFONXPJCBIS-UHFFFAOYNA-N
Synonym phenyl pyridin-2-yl methanamine hydrochloride, 1-phenyl-1-pyridin-2-ylmethanamine hydrochloride, phenyl 2-pyridyl methylamine hydrochloride, phenyl-2-pyridylmethylamine, chloride, phenyl 2-pyridyl methanamine hydrochloride, phenyl pyridin-2-yl methylamine hydrochloride, alpha-pyridin-2-yl benzylamine hydrochloride, phenyl 2-pyridyl methylamine dihydrochloride, 1-phenyl-1-pyridin-2-yl methanamine hydrochloride
PubChem CID 2775257
IUPAC Name phenyl(pyridin-2-yl)methanamine;hydrochloride
SMILES Cl.NC(C1=CC=CC=C1)C1=CC=CC=N1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 257.16
Percent Purity 95%
Chemical Name or Material 1-Phenyl-1-pyridin-2-ylmethanamine dihydrochloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'