missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Phenyl-2-pyrrolidinone, 99%, Thermo Scientific Chemicals
£26.40 - £204.00
Chemical Identifiers
CAS | 4641-57-0 |
---|---|
Molecular Formula | C10H11NO |
Molecular Weight (g/mol) | 161.20 |
MDL Number | MFCD00003192 |
InChI Key | JMVIVASFFKKFQK-UHFFFAOYSA-N |
Synonym | 1-phenyl-2-pyrrolidinone, 2-pyrrolidinone, 1-phenyl, n-phenyl-2-pyrrolidone, n-phenyl-2-pyrrolidinone, 1-phenyl-pyrrolidin-2-one, 1-phenyl-2-pyrrolidone, n-phenylpyrrolidinone, n-phenyl pyrrolidone, pubchem23691, maybridge3_004960 |
PubChem CID | 78375 |
IUPAC Name | 1-phenylpyrrolidin-2-one |
SMILES | O=C1CCCN1C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11373636
|
Thermo Scientific Alfa Aesar
L14660.06 |
5 g |
£26.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11383636
|
Thermo Scientific Alfa Aesar
L14660.14 |
25 g |
£83.80
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11393636
|
Thermo Scientific Alfa Aesar
L14660.22 |
100 g |
£204.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
A novel derivative of 1-phenyl-2-pyrrolidinone, blebbistatin, inhibits non-muscle myocin II activity with high specificity.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsA novel derivative of 1-phenyl-2-pyrrolidinone, blebbistatin, inhibits non-muscle myocin II activity with high specificity.
Solubility
Soluble in chloroform and ethyl Acetate. Insoluble in water.
Notes
Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
4641-57-0 | |
161.20 | |
JMVIVASFFKKFQK-UHFFFAOYSA-N | |
78375 | |
O=C1CCCN1C1=CC=CC=C1 |
C10H11NO | |
MFCD00003192 | |
1-phenyl-2-pyrrolidinone, 2-pyrrolidinone, 1-phenyl, n-phenyl-2-pyrrolidone, n-phenyl-2-pyrrolidinone, 1-phenyl-pyrrolidin-2-one, 1-phenyl-2-pyrrolidone, n-phenylpyrrolidinone, n-phenyl pyrrolidone, pubchem23691, maybridge3_004960 | |
1-phenylpyrrolidin-2-one |
Specifications
4641-57-0 | |
123°C (0.2 mmHg) | |
C10H11NO | |
130685 | |
Soluble in chloroform and ethyl Acetate. Insoluble in water. | |
O=C1CCCN1C1=CC=CC=C1 | |
161.20 | |
161.2 | |
1-Phenyl-2-pyrrolidinone |
63°C to 68°C | |
Odorless | |
MFCD00003192 | |
1-phenyl-2-pyrrolidinone, 2-pyrrolidinone, 1-phenyl, n-phenyl-2-pyrrolidone, n-phenyl-2-pyrrolidinone, 1-phenyl-pyrrolidin-2-one, 1-phenyl-2-pyrrolidone, n-phenylpyrrolidinone, n-phenyl pyrrolidone, pubchem23691, maybridge3_004960 | |
JMVIVASFFKKFQK-UHFFFAOYSA-N | |
1-phenylpyrrolidin-2-one | |
78375 | |
99% |
Safety and Handling
missing translation for 'einecsNumber' : 225-069-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only