Learn More
1-Phenyl-3-trimethylsilyl-2-propyn-1-ol, 97%, Thermo Scientific Chemicals
£47.03 - £173.87
Chemical Identifiers
CAS | 89530-34-7 |
---|---|
Molecular Formula | C12H16OSi |
Molecular Weight (g/mol) | 204.344 |
MDL Number | MFCD05864336 |
InChI Key | RVTDALNDYLDVMN-UHFFFAOYSA-N |
Synonym | 1-phenyl-3-trimethylsilyl-2-propyn-1-ol, 1-phenyl-3-trimethylsilyl prop-2-yn-1-ol, benzenemethanol, a-2-trimethylsilyl ethynyl, acmc-20ln8h, alpha-trimethylsilyl ethynylbenzyl alcohol, 4,4-dimethyl-1-phenyl-4-silapent-2-yn-1-ol, alpha-trimethylsilyl ethynyl-benzenemethanol, alpha-trimethylsilyl-ethynyl-benzenemethanol, alpha-trimethylsilyl ethynyl-benzene-methanol, 1-phenyl-3-1,1,1-trimethylsilyl-2-propyn-1-olShow More |
PubChem CID | 2760416 |
IUPAC Name | 1-phenyl-3-trimethylsilylprop-2-yn-1-ol |
SMILES | C[Si](C)(C)C#CC(C1=CC=CC=C1)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15430228
|
Thermo Scientific Alfa Aesar
H53426.06 |
5 g |
£47.03
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15440228
|
Thermo Scientific Alfa Aesar
H53426.14 |
25 g |
£173.87
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
89530-34-7 | |
204.344 | |
RVTDALNDYLDVMN-UHFFFAOYSA-N | |
2760416 | |
C[Si](C)(C)C#CC(C1=CC=CC=C1)O |
C12H16OSi | |
MFCD05864336 | |
1-phenyl-3-trimethylsilyl-2-propyn-1-ol, 1-phenyl-3-trimethylsilyl prop-2-yn-1-ol, benzenemethanol, a-2-trimethylsilyl ethynyl, acmc-20ln8h, alpha-trimethylsilyl ethynylbenzyl alcohol, 4,4-dimethyl-1-phenyl-4-silapent-2-yn-1-ol, alpha-trimethylsilyl ethynyl-benzenemethanol, alpha-trimethylsilyl-ethynyl-benzenemethanol, alpha-trimethylsilyl ethynyl-benzene-methanol, 1-phenyl-3-1,1,1-trimethylsilyl-2-propyn-1-olShow More | |
1-phenyl-3-trimethylsilylprop-2-yn-1-ol |
Specifications
89530-34-7 | |
MFCD05864336 | |
1-phenyl-3-trimethylsilyl-2-propyn-1-ol, 1-phenyl-3-trimethylsilyl prop-2-yn-1-ol, benzenemethanol, a-2-trimethylsilyl ethynyl, acmc-20ln8h, alpha-trimethylsilyl ethynylbenzyl alcohol, 4,4-dimethyl-1-phenyl-4-silapent-2-yn-1-ol, alpha-trimethylsilyl ethynyl-benzenemethanol, alpha-trimethylsilyl-ethynyl-benzenemethanol, alpha-trimethylsilyl ethynyl-benzene-methanol, 1-phenyl-3-1,1,1-trimethylsilyl-2-propyn-1-ol | |
C[Si](C)(C)C#CC(C1=CC=CC=C1)O | |
204.344 | |
204.34 | |
1.5193 | |
1-Phenyl-3-trimethylsilyl-2-propyn-1-ol |
C12H16OSi | |
110°C (230°F) | |
RVTDALNDYLDVMN-UHFFFAOYSA-N | |
1-phenyl-3-trimethylsilylprop-2-yn-1-ol | |
2760416 | |
97% | |
0.961 |
Safety and Handling
GHS H Statement
H319
Causes serious eye irritation.
P264b-P280i-P305+P351+P338
H319
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.